Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146161.1 | NP_001139633.1 | 10 - 11 | 1120 | 98 | |
NM_008732.2 | NP_032758.2 | 10 - 11 | 1120 | 98 | ||
GenBank mRNA | AF029758.1 | - | 10 - 11 | 1055 | 98 | |
AK049856.1 | - | 10 - 11 | 1068 | 98 | ||
AK083478.1 | - | 10 - 11 | 1066 | 98 | ||
AK148276.1 | - | 10 - 11 | 1068 | 98 | ||
BC019137.1 | - | 10 - 11 | 1094 | 5040976 | 98 | |
L33415.1 | - | 10 - 11 | 1048 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |