Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081030.2 | NP_001074499.2 | 30 - 31 | 4379 | 59 | |
NM_001170561.1 | NP_001164032.1 | 31 - 32 | 4457 | 59 | ||
GenBank mRNA | AK129485.1 | - | 30 - 31 | 4323 | 59 | |
AK147332.1 | - | 30 - 31 | 4336 | 59 | ||
BC098209.1 | - | 11 - 12 | 1722 | 59 | ||
BC157935.1 | - | 30 - 31 | 4269 | 9088115 | 59 | |
BC158013.1 | - | 31 - 32 | 4347 | 9007320 | 59 | |
BC172094.1 | - | 30 - 31 | 4269 | 9054380 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |