Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048178.2 | NP_001041643.1 | 2 - 3 | 429 | 91 | |
NM_001286370.1 | NP_001273299.1 | 2 - 3 | 130 | 91 | ||
NM_010284.3 | NP_034414.2 | 2 - 3 | 429 | 91 | ||
GenBank mRNA | AK053579.1 | - | 2 - 3 | 290 | 91 | |
AK143886.1 | - | 2 - 3 | 131 | 91 | ||
AK143925.1 | - | 2 - 3 | 227 | 91 | ||
AK148440.1 | - | 2 - 3 | 515 | 91 | ||
BC024375.1 | - | 2 - 3 | 264 | 91 | ||
BC056645.1 | - | 2 - 3 | 245 | 91 | ||
BC075720.1 | - | 2 - 3 | 277 | 5044060 | 91 | |
BY754177.1 | - | 2 - 3 | 430 | 91 | ||
M31680.1 | - | 2 - 3 | 165 | 91 | ||
M33324.1 | - | 2 - 3 | 165 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |