Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048178.2 | NP_001041643.1 | 4 - 5 | 629 | 78 | |
NM_001286370.1 | NP_001273299.1 | 4 - 5 | 330 | 78 | ||
NM_010284.3 | NP_034414.2 | 4 - 5 | 629 | 78 | ||
GenBank mRNA | AK143886.1 | - | 4 - 5 | 331 | 78 | |
AK143925.1 | - | 4 - 5 | 427 | 78 | ||
BC024375.1 | - | 4 - 5 | 464 | 78 | ||
BC056645.1 | - | 4 - 5 | 445 | 78 | ||
BC075720.1 | - | 4 - 5 | 477 | 5044060 | 78 | |
M31680.1 | - | 4 - 5 | 365 | 78 | ||
M33324.1 | - | 4 - 5 | 365 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |