Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198927.3 | NP_945121.1 | 14 - 15 | 1633 | 75 | |
GenBank mRNA | AK132579.1 | - | 13 - 14 | 1536 | 75 | |
AY459348.1 | - | 14 - 15 | 1634 | 75 | ||
BC038603.1 | - | 4 - 5 | 389 | 75 | ||
BC049243.1 | - | 6 - 7 | 638 | 75 | ||
BC069958.1 | - | 5 - 6 | 465 | 75 | ||
BC138091.1 | - | 14 - 15 | 1634 | 8861110 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |