Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018814.3 | NP_061284.2 | 34 - 35 | 7089 | 101 | |
GenBank mRNA | AF096286.1 | - | 28 - 29 | 4919 | 101 | |
AK014387.1 | - | 8 - 9 | 1777 | 101 | ||
AK129221.1 | - | 29 - 30 | 4969 | 101 | ||
AK154122.1 | - | 15 - 16 | 2817 | 101 | ||
AK170320.1 | - | 15 - 16 | 2817 | 101 | ||
BC066849.1 | - | 29 - 30 | 4752 | 101 | ||
BC098370.1 | - | 29 - 30 | 4679 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |