Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_001165952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_001165952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_001165953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_001165953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_194343.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
NM_194343.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008148.mm.1 |
TC0300001007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_001165952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_001165952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_001165953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_001165953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_194343.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
NM_194343.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008149.mm.1 |
TC0300001007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_001165952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_001165952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_001165953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_001165953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_194343.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
NM_194343.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008150.mm.1 |
TC0300001007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_001165952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_001165952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_001165953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_001165953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_194343.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
NM_194343.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008147.mm.1 |
TC0300001007.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_001165952.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_001165952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_001165953.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_001165953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_194343.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
NM_194343.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008151.mm.1 |
TC0300001007.mm.1 |
MTA TC |