Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053180.2 | NP_444410.1 | 4 - 5 | 541 | 66 | |
GenBank mRNA | AY005133.1 | - | 4 - 5 | 552 | 66 | |
BC031907.1 | - | 4 - 5 | 541 | 5362198 | 66 | |
BC058353.1 | - | 822 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |