Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033177.2 | NP_001028349.1 | 2 - 3 | 830 | 66 | |
GenBank mRNA | AK009075.1 | - | 2 - 3 | 813 | 66 | |
AK036802.1 | - | 2 - 3 | 831 | 66 | ||
AK137676.1 | - | 2 - 3 | 831 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |