Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spryd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033277.3 | NP_001028449.2 | 8 - 9 | 959 | 55 | |
GenBank mRNA | AK146689.1 | - | 8 - 9 | 962 | 55 | |
BC008150.1 | - | 4 - 5 | 470 | 55 | ||
BC048820.1 | - | 7 - 8 | 1188 | 55 | ||
BG293312.1 | - | 1 - 2 | 44 | 55 | ||
CX201967.1 | - | 4 - 5 | 458 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |