Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029788.5 | NP_084064.1 | 5 - 6 | 915 | 138 | |
GenBank mRNA | AK002624.1 | - | 5 - 6 | 873 | 138 | |
AK036401.1 | - | 5 - 6 | 916 | 138 | ||
AK153293.1 | - | 5 - 6 | 884 | 138 | ||
AK171077.1 | - | 4 - 5 | 2166 | 138 | ||
BC037057.1 | - | 4 - 5 | 702 | 138 | ||
BC050796.1 | - | 5 - 6 | 883 | 6743413 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |