Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144812.2 | NP_659061.2 | 11 - 12 | 3611 | 70 | |
NM_177124.4 | NP_796098.3 | 8 - 9 | 3402 | 70 | ||
GenBank mRNA | AK052247.1 | - | 3 - 4 | 365 | 70 | |
AK162702.1 | - | 2 - 3 | 102 | 70 | ||
AK162788.1 | - | 3 - 4 | 658 | 70 | ||
BC068158.1 | - | 8 - 9 | 3297 | 70 | ||
BC115729.1 | - | 4 - 5 | 695 | 70 | ||
CN704499.1 | - | 4 - 5 | 494 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |