Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00008657.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008657.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008657.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008651.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008651.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008651.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008656.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008656.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008656.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008661.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008661.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008661.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008653.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008653.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008653.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008646.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008646.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008646.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008650.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008650.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008650.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008652.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008652.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008652.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008655.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008655.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008655.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008644.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008644.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008644.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008645.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008645.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008645.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008648.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008648.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008648.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008659.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008659.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008659.mm.1 |
TC0X00001372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008658.mm.1 |
NM_001081499.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008658.mm.1 |
NM_001081499.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008658.mm.1 |
TC0X00001372.mm.1 |
MTA TC |