Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289603.1 | NP_001276532.1 | 9 - 10 | 1043 | 72 | |
NM_033476.3 | NP_258437.1 | 9 - 10 | 1043 | 72 | ||
GenBank mRNA | AK146503.1 | - | 9 - 10 | 1044 | 72 | |
AK147898.1 | - | 8 - 9 | 1252 | 72 | ||
AK158682.1 | - | 9 - 10 | 963 | 72 | ||
BC022131.1 | - | 9 - 10 | 1016 | 5098336 | 72 | |
M84809.1 | - | 9 - 10 | 982 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |