Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033218.1 | NP_150087.1 | 13 - 14 | 2596 | 86 | |
GenBank mRNA | AF374267.1 | - | 13 - 14 | 2449 | 86 | |
AK155857.1 | - | 14 - 15 | 2544 | 86 | ||
AK172219.1 | - | 13 - 14 | 2604 | 86 | ||
AK183936.1 | - | 1 - 2 | 64 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |