Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | ||||
GenBank mRNA | AK035870.1 | - | ||||
AK043769.1 | - | |||||
AK137169.1 | - |