Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028918.2 | NP_083194.2 | 4 - 5 | 629 | 71 | |
GenBank mRNA | AK016657.1 | - | 4 - 5 | 560 | 71 | |
AK161541.1 | - | 4 - 5 | 568 | 71 | ||
AK162855.1 | - | 4 - 5 | 569 | 71 | ||
BC050805.1 | - | 4 - 5 | 653 | 6771545 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |