Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009510.2 | NP_033536.2 | 2 - 3 | 230 | 55 | |
GenBank mRNA | AK002766.1 | - | 2 - 3 | 204 | 55 | |
AK004588.2 | - | 2 - 3 | 205 | 55 | ||
AK135194.1 | - | 2 - 3 | 206 | 55 | ||
AK135353.1 | - | 2 - 3 | 206 | 55 | ||
AK145707.1 | - | 2 - 3 | 231 | 55 | ||
AK159717.1 | - | 2 - 3 | 208 | 55 | ||
AK159824.1 | - | 2 - 3 | 206 | 55 | ||
AK159937.1 | - | 2 - 3 | 207 | 55 | ||
AK166191.1 | - | 2 - 3 | 206 | 55 | ||
AK168410.1 | - | 2 - 3 | 206 | 55 | ||
AK168840.1 | - | 2 - 3 | 206 | 55 | ||
AK170610.1 | - | 2 - 3 | 206 | 55 | ||
AK172336.1 | - | 2 - 3 | 213 | 55 | ||
BC048181.1 | - | 2 - 3 | 193 | 6826190 | 55 | |
BC098502.1 | - | 2 - 3 | 182 | 30047137 | 55 | |
X60671.1 | - | 2 - 3 | 188 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |