Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029786.1 | - | 8 - 9 | 2556 | 95 | |
AK132195.1 | - | 8 - 9 | 3964 | 95 | ||
AK162464.1 | - | 7 - 8 | 2631 | 95 | ||
BC072599.1 | - | 8 - 9 | 3975 | 95 | ||
BC079861.1 | - | 8 - 9 | 3962 | 6834767 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |