Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009705.3 | NP_033835.1 | 1 - 2 | 196 | 79 | |
GenBank mRNA | AF032466.1 | - | 1 - 2 | 148 | 79 | |
BC023349.1 | - | 1 - 2 | 160 | 5254031 | 79 | |
DV039756.1 | - | 1 - 2 | 230 | 79 | ||
U90886.1 | - | 1 - 2 | 168 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |