Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033425.4 | NP_001028597.2 | 3 - 4 | 859 | 96 | |
NM_001289481.1 | NP_001276410.1 | 3 - 4 | 859 | 96 | ||
NM_001289483.1 | NP_001276412.1 | 3 - 4 | 355 | 96 | ||
NM_001289484.1 | NP_001276413.1 | 3 - 4 | 724 | 96 | ||
GenBank mRNA | AK141259.1 | - | 3 - 4 | 830 | 96 | |
BC145638.1 | - | 2 - 3 | 692 | 9054214 | 96 | |
DQ323929.1 | - | 3 - 4 | 827 | 96 | ||
DQ323930.1 | - | 3 - 4 | 827 | 96 | ||
EF152498.1 | - | 2 - 3 | 674 | 96 | ||
EF152499.1 | - | 3 - 4 | 319 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |