Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011767.2 | NP_035897.2 | 17 - 18 | 2900 | 78 | |
GenBank mRNA | AF071059.1 | - | 17 - 18 | 2898 | 78 | |
AK011329.1 | - | 2 - 3 | 220 | 78 | ||
AK034963.1 | - | 10 - 11 | 1469 | 78 | ||
AK158938.1 | - | 9 - 10 | 1208 | 78 | ||
BC006962.1 | - | 7 - 8 | 947 | 78 | ||
BC094220.1 | - | 16 - 17 | 2787 | 78 | ||
BC141139.1 | - | 17 - 18 | 2884 | 9055751 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |