Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161628.1 | NP_001155100.1 | 12 - 13 | 2113 | 107 | |
NM_001161629.1 | NP_001155101.1 | 10 - 11 | 1601 | 107 | ||
NM_029946.4 | NP_084222.4 | 29 - 30 | 4220 | 107 | ||
GenBank mRNA | AK016516.1 | - | 10 - 11 | 1706 | 107 | |
AK019850.1 | - | 12 - 13 | 2113 | 107 | ||
AK163363.1 | - | 7 - 8 | 1291 | 107 | ||
AK220412.1 | - | 12 - 13 | 2032 | 107 | ||
BC056955.1 | - | 17 - 18 | 2623 | 6812632 | 107 | |
BC065111.1 | - | 29 - 30 | 4220 | 30536704 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |