Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080420.2 | NP_536345.2 | 11 - 12 | 2059 | 63 | |
GenBank mRNA | AK162992.1 | - | 3 - 4 | 507 | 63 | |
BC016212.1 | - | 11 - 12 | 2066 | 4924899 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |