Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013749.2 | NP_038777.2 | 2 - 3 | 230 | 68 | |
GenBank mRNA | AF156164.1 | - | 2 - 3 | 225 | 68 | |
AK005382.1 | - | 2 - 3 | 227 | 68 | ||
AK005530.1 | - | 2 - 3 | 226 | 68 | ||
AK160136.1 | - | 2 - 3 | 231 | 68 | ||
BC025860.1 | - | 2 - 3 | 214 | 5251646 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |