Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030224.1 | NP_084500.1 | 2 - 3 | 628 | 78 | |
GenBank mRNA | AK019639.1 | - | 2 - 3 | 650 | 78 | |
AK085347.1 | - | 1 - 2 | 386 | 78 | ||
BC120835.1 | - | 2 - 3 | 628 | 40129758 | 78 | |
BC137682.1 | - | 2 - 3 | 628 | 8860701 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |