Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276402.1 | NP_001263331.1 | 1 - 2 | 1383 | 91 | |
NM_010703.4 | NP_034833.2 | 1 - 2 | 1383 | 91 | ||
GenBank mRNA | AK028246.1 | - | 1 - 2 | 1005 | 91 | |
AK034819.1 | - | 1 - 2 | 856 | 91 | ||
AK131979.1 | - | 1 - 2 | 1365 | 91 | ||
AK158769.1 | - | 1 - 2 | 1365 | 91 | ||
BC057543.1 | - | 1 - 2 | 1389 | 6401514 | 91 | |
X58636.1 | - | 1 - 2 | 1197 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |