Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028943.5 | NP_083219.2 | 2 - 3 | 1023 | 120 | |
GenBank mRNA | AK016659.1 | - | 2 - 3 | 1037 | 120 | |
BC117782.2 | - | 2 - 3 | 1027 | 40092081 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |