Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300023381.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023381.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023381.mm.1 |
TC0300002957.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023383.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023383.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023383.mm.1 |
TC0300002957.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023385.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023385.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023385.mm.1 |
TC0300002957.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023384.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023384.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023384.mm.1 |
TC0300002957.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023382.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023382.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023382.mm.1 |
TC0300002957.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300023386.mm.1 |
NM_027816.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023386.mm.1 |
NM_027816.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300023386.mm.1 |
TC0300002957.mm.1 |
MTA TC |