Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014763.1 | - | 1 - 2 | 343 | 89 | |
AK032034.1 | - | 1 - 2 | 709 | 89 | ||
AK076176.1 | - | 1 - 2 | 473 | 89 | ||
AK082692.1 | - | 1 - 2 | 605 | 89 | ||
AY512657.1 | - | 1 - 2 | 256 | 89 | ||
BB611685.1 | - | 1 - 2 | 473 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons |