Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400000973.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000973.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000975.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000975.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000977.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000977.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000994.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000994.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001002.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001002.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000979.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000979.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001000.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001000.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001003.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001003.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000976.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000976.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000982.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000982.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000992.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000992.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000967.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000967.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000968.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000968.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000981.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000981.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000984.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000984.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000988.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000988.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001001.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001001.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000972.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000972.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000986.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000986.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001006.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001006.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000966.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000966.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000969.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000969.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000980.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000980.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000989.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000989.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400001004.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400001004.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000970.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000970.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000971.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000971.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000974.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000974.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000978.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000978.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000990.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000990.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000993.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000993.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000983.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000983.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000987.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000987.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000991.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000991.mm.1 |
TC1400000120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000995.mm.1 |
NM_001195229.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000995.mm.1 |
TC1400000120.mm.1 |
MTA TC |