Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172597.2 | NP_766185.1 | 14 - 15 | 1630 | 144 | |
GenBank mRNA | AK046466.1 | - | 14 - 15 | 1631 | 144 | |
AK173153.1 | - | 2127 | 144 | |||
BC052488.1 | - | 14 - 15 | 1506 | 6438726 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |