Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130890.2 | NP_570960.2 | 14 - 15 | 1690 | 64 | |
GenBank mRNA | AB050202.1 | - | 14 - 15 | 1686 | 64 | |
AB061518.1 | - | 14 - 15 | 1636 | 64 | ||
BC125504.1 | - | 14 - 15 | 1662 | 40130019 | 64 | |
BC125508.1 | - | 14 - 15 | 1662 | 40130023 | 64 | |
BC145055.1 | - | 14 - 15 | 1662 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |