Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008882.2 | NP_032908.2 | 30 - 31 | 6076 | 73 | |
GenBank mRNA | AK122289.1 | - | 29 - 30 | 4340 | 73 | |
BC024509.1 | - | 5 - 6 | 736 | 73 | ||
BC051045.1 | - | 4 - 5 | 862 | 73 | ||
BC056475.1 | - | 10 - 11 | 1527 | 73 | ||
BC068155.1 | - | 30 - 31 | 5858 | 30362726 | 73 | |
D86949.1 | - | 30 - 31 | 5982 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |