Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103182.2 | NP_001096652.2 | 5 - 6 | 620 | 127 | |
NR_120498.1 | 5 - 6 | 620 | 127 | |||
GenBank mRNA | AF190325.1 | - | 5 - 6 | 436 | 127 | |
AK042357.1 | - | 5 - 6 | 402 | 127 | ||
AK052618.1 | - | 5 - 6 | 418 | 127 | ||
AK089989.1 | - | 5 - 6 | 621 | 127 | ||
BC069181.1 | - | 2 - 3 | 159 | 127 | ||
BC088992.1 | - | 5 - 6 | 410 | 127 | ||
BC099695.1 | - | 5 - 6 | 404 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |