Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172742.2 | NP_766330.2 | 1 - 2 | 169 | 95 | |
GenBank mRNA | BC041672.1 | - | 1 - 2 | 141 | 95 | |
CK634659.1 | - | 1 - 2 | 55 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |