Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207636.2 | NP_997519.2 | 6 - 7 | 1079 | 91 | |
GenBank mRNA | AK032733.1 | - | 6 - 7 | 1051 | 91 | |
AK048586.1 | - | 6 - 7 | 1082 | 91 | ||
AK089380.1 | - | 6 - 7 | 1044 | 91 | ||
AK154790.1 | - | 6 - 7 | 848 | 91 | ||
BC144880.1 | - | 6 - 7 | 887 | 91 | ||
DQ192036.1 | - | 6 - 7 | 1080 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |