Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040690.2 | NP_001035780.1 | 11 - 12 | 1461 | 76 | |
NM_145544.3 | NP_663519.2 | 10 - 11 | 1314 | 76 | ||
GenBank mRNA | AK052125.1 | - | 10 - 11 | 1262 | 76 | |
AK154852.1 | - | 11 - 12 | 1447 | 76 | ||
AK155793.1 | - | 11 - 12 | 1447 | 76 | ||
AK164159.1 | - | 11 - 12 | 1373 | 76 | ||
AK166315.1 | - | 11 - 12 | 1394 | 76 | ||
AK172052.1 | - | 11 - 12 | 1463 | 76 | ||
BC011279.1 | - | 10 - 11 | 1183 | 4008751 | 76 | |
CF731381.1 | - | 1 - 2 | 89 | 76 | ||
CN702379.1 | - | 2 - 3 | 274 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |