Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166648.1 | NP_001160120.1 | 13 - 14 | 1561 | 103 | |
NM_001166649.1 | NP_001160121.1 | 13 - 14 | 1582 | 103 | ||
NM_001166650.1 | NP_001160122.1 | 12 - 13 | 1520 | 103 | ||
NM_153532.3 | NP_705760.2 | 12 - 13 | 1892 | 103 | ||
GenBank mRNA | AK129399.1 | - | 13 - 14 | 1541 | 103 | |
AK141491.1 | - | 13 - 14 | 1562 | 103 | ||
BC028839.1 | - | 12 - 13 | 1532 | 5150255 | 103 | |
BC051397.1 | - | 12 - 13 | 1520 | 5150255 | 103 | |
BC063775.1 | - | 12 - 13 | 1904 | 5375514 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |