Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286482.1 | NP_001273411.1 | 3 - 4 | 510 | 68 | |
NM_001286483.1 | NP_001273412.1 | 2 - 3 | 350 | 68 | ||
NM_144841.4 | NP_659090.1 | 1 - 2 | 432 | 68 | ||
GenBank mRNA | AK053741.1 | - | 2 - 3 | 347 | 68 | |
AK081535.1 | - | 2 - 3 | 351 | 68 | ||
AK087527.1 | - | 2 - 3 | 348 | 68 | ||
BC017609.1 | - | 1 - 2 | 432 | 4527414 | 68 | |
BQ959742.1 | - | 3 - 4 | 510 | 68 | ||
CO424557.1 | - | 3 - 4 | 435 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |