Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027288.3 | NP_081564.3 | 6 - 7 | 924 | 118 | |
GenBank mRNA | AF306557.1 | - | 6 - 7 | 928 | 118 | |
AK036505.1 | - | 7 - 8 | 1024 | 118 | ||
AK047057.1 | - | 6 - 7 | 917 | 118 | ||
AK142165.1 | - | 6 - 7 | 889 | 118 | ||
AK154172.1 | - | 6 - 7 | 905 | 119 | ||
AK166022.1 | - | 6 - 7 | 119 | |||
AK170290.1 | - | 6 - 7 | 921 | 118 | ||
BC031409.1 | - | 6 - 7 | 867 | 4952710 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |