Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019671.1 | - | 5 - 6 | 1402 | 79 | |
BC034362.1 | - | 4 - 5 | 1147 | 3154701 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |