Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253759.1 | NP_001240688.1 | 10 - 11 | 1575 | 60 | |
NM_172813.3 | NP_766401.1 | 10 - 11 | 1575 | 60 | ||
GenBank mRNA | AK045500.1 | - | 10 - 11 | 1493 | 60 | |
AK051821.1 | - | 10 - 11 | 1523 | 60 | ||
AK082388.1 | - | 10 - 11 | 1576 | 60 | ||
AK162001.1 | - | 10 - 11 | 1524 | 60 | ||
BC052062.1 | - | 7 - 8 | 1086 | 5703279 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |