Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il3ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008369.1 | NP_032395.1 | 5 - 6 | 607 | 64 | |
GenBank mRNA | BC049889.1 | - | 4 - 5 | 1007 | 64 | |
FJ550346.1 | - | 2 - 3 | 189 | 64 | ||
X64534.1 | - | 5 - 6 | 607 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |