Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193659.1 | NP_001180588.1 | 2 - 3 | 313 | 72 | |
NM_001193660.1 | NP_001180589.1 | 3 - 4 | 261 | 72 | ||
NM_001193661.1 | NP_001180590.1 | 2 - 3 | 148 | 72 | ||
NM_022890.2 | NP_075028.1 | 2 - 3 | 303 | 72 | ||
GenBank mRNA | AK039672.1 | - | 2 - 3 | 195 | 72 | |
AK049331.1 | - | 2 - 3 | 170 | 72 | ||
AK128999.1 | - | 2 - 3 | 182 | 72 | ||
AK132155.1 | - | 2 - 3 | 172 | 72 | ||
BC024057.1 | - | 2 - 3 | 212 | 5100104 | 72 | |
BY012969.1 | - | 3 - 4 | 261 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |