Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028938.2 | NP_083214.2 | 2 - 3 | 482 | 141 | |
GenBank mRNA | AK015333.1 | - | 2 - 3 | 379 | 141 | |
AK015758.1 | - | 2 - 3 | 329 | 141 | ||
AK016634.1 | - | 2 - 3 | 377 | 141 | ||
AK087661.1 | - | 2 - 3 | 482 | 141 | ||
AK132656.1 | - | 2 - 3 | 447 | 141 | ||
BC112424.1 | - | 2 - 3 | 339 | 40062088 | 141 | |
BC113130.1 | - | 2 - 3 | 339 | 40062093 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |