Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010937.2 | NP_001010937.1 | 2 - 3 | 592 | 92 | |
GenBank mRNA | AK134617.1 | - | 2 - 3 | 591 | 92 | |
AK141610.1 | - | 2 - 3 | 592 | 92 | ||
BC013811.1 | - | 2 - 3 | 259 | 3483477 | 92 | |
BC016507.1 | - | 2 - 3 | 267 | 3481872 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |