Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045489.1 | NP_001038954.1 | 4 - 5 | 603 | 71 | |
NM_008594.2 | NP_032620.2 | 5 - 6 | 714 | 71 | ||
GenBank mRNA | AB021130.2 | - | 5 - 6 | 621 | 71 | |
AB025280.1 | - | 4 - 5 | 510 | 71 | ||
AK087555.1 | - | 4 - 5 | 593 | 71 | ||
AK089211.1 | - | 4 - 5 | 569 | 71 | ||
AK145183.1 | - | 4 - 5 | 567 | 71 | ||
AK148371.1 | - | 4 - 5 | 592 | 71 | ||
AK152088.1 | - | 5 - 6 | 703 | 71 | ||
AK155047.1 | - | 5 - 6 | 712 | 71 | ||
AK169997.1 | - | 4 - 5 | 567 | 71 | ||
AK170935.1 | - | 4 - 5 | 567 | 71 | ||
AK171143.1 | - | 5 - 6 | 678 | 71 | ||
BC003892.1 | - | 4 - 5 | 547 | 3593310 | 71 | |
BC003904.1 | - | 4 - 5 | 542 | 3601565 | 71 | |
BC018577.1 | - | 4 - 5 | 540 | 71 | ||
BC037475.1 | - | 1 - 2 | 929 | 71 | ||
Y11684.2 | - | 4 - 5 | 510 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |