Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286750.1 | NP_001273679.1 | 3 - 4 | 367 | 71 | |
NM_153157.3 | NP_694797.1 | 3 - 4 | 518 | 71 | ||
NM_153458.3 | NP_703188.1 | 3 - 4 | 771 | 71 | ||
GenBank mRNA | AF442824.1 | - | 3 - 4 | 518 | 71 | |
AF442825.1 | - | 3 - 4 | 486 | 71 | ||
AK038569.1 | - | 3 - 4 | 718 | 71 | ||
AK044325.1 | - | 3 - 4 | 573 | 71 | ||
AK044605.1 | - | 3 - 4 | 721 | 71 | ||
AK045482.1 | - | 3 - 4 | 720 | 71 | ||
AK049612.1 | - | 3 - 4 | 772 | 71 | ||
AK079315.1 | - | 3 - 4 | 581 | 71 | ||
AK137414.1 | - | 3 - 4 | 573 | 71 | ||
BB628879.1 | - | 3 - 4 | 368 | 71 | ||
BC112422.1 | - | 3 - 4 | 427 | 40061966 | 71 | |
BC113129.1 | - | 3 - 4 | 427 | 40061967 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |