Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291222.1 | NP_001278151.1 | 14 - 15 | 3151 | 81 | |
NM_010626.3 | NP_034756.2 | 14 - 15 | 3148 | 81 | ||
GenBank mRNA | BC034093.1 | - | 4 - 5 | 723 | 81 | |
BC141045.1 | - | 14 - 15 | 3149 | 9055657 | 81 | |
BC145222.1 | - | 14 - 15 | 3152 | 9053773 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |